Learn More
Corning™ Bottle Top Dispenser (BTD) PTFE Plunger
PTFE plunger for BTD and dispensing reagents
Brand:  Corning™ 6865
 View more versions of this product
Product Code. 16842110
  • 849.56 SEK / Each
Only null left
Estimated Shipment: 30-10-2025
Description
  • Specialized airpurging and closed circuit system ensures no loss of reagent during dispensing, as well as when the device is not in use
  • Comes with four adapters with different diameters for universal use with multiple bottle sizes
  • Telescopic suction tube can be adjusted smoothly to different bottle heights
  • Calibration tool allows easy in-lab calibration with built-in recalibration mechanism to ensure the highest reproducibility during the dispenser’s long life cycle
  • Every dispenser is individually tested and delivered with a certificate of performance
  • Uses:
    • For dispensing organic and inorganic liquids
    TRUSTED_SUSTAINABILITY
  • Specifications
    Specifications
    50 mL
    Dispensing Organic and Inorganic Liquids
    Four adapters
    Documents
    One moment while we fetch your results.
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Corning™ Bottle Top Dispenser (BTD) PTFE Plunger > 50 mL