Learn More
Trajan™ BP1 GC Capillary Columns
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
Brand:  Trajan™ 054070
 View more versions of this product
Product Code. 12653016
  • 5030.00 SEK / Each
Only null left
Estimated Shipment: 23-07-2025
Description
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Specifications
340/360°C
0.32 mm
G1, G2, G38
-60°C to +340/360°C
12 m
Non polar
Van der Waals interaction
100% Dimethyl Polysiloxane
Amines Analysis, Aromatics Analysis, Pesticide Analysis, Phenols, Herbicides Analysis, Amines
1 ÎĽm
Forte™ BP1
Fused Silica
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Trajan™ BP1 GC Capillary Columns > I.D.: 0.32mm; Length: 12m; Film Thickness: 1um