Learn More
Trajan Scientific and Medical™ BPX50 GC Capillary Columns
Designed for range of EPA and pharmaceutical methods. Trajan Scientific and Medical BPX50 GC Capillary Columns are MS premium, low bleed phase columns.
Brand: Trajan Scientific and Medical™ 054742
Additional Details : Weight : 0.01000kg
Description
- Mid-polarity column
- Inert
- Low bleed
- High temperature
- Ideal for a range of EPA methods and pharmaceutical applications
Specifications
BPX50 GC Capillary Column | |
Triazines/Herbicides Analysis, Triazines/Herbicides, Drug Screening, Steroids and a Variety of Pharmaceutical Applications | |
0.18 μm | |
50% Phenyl Polysilphenylene-siloxane |
0.18 mm | |
80°C to 330/350°C | |
20 m | |
BPX50 |