Learn More
VACUUBRAND™ Rotary Vane Vacuum Pumps: Oil Mist Filters
Oil Mist Filters and replacements for VACUUBRAND Rotary Vane Vacuum Pumps
Brand:  VACUUBRAND™
Additional Details : Weight : 0.81540kg
 View more versions of this product
Product Code. 10292551
  • 4515.00 SEK / Each
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
  • For use with VACUUBRAND Rotary Vane Vacuum Pumps
  • Oil Mist Filters “FO” trap up to 99% of oil mist generated by the pump, helping preserve the laboratory environment
Specifications
Specifications
Model RE2, RZ2, RE2.5, RZ2.5, RE5, RZ5, RE6, RZ6 rotary vane vacuum pump
Oil Mist Filter