Learn More
VACUUBRAND™ Rotary Vane Vacuum Pumps: Oil Mist Filters
Oil Mist Filters and replacements for VACUUBRAND Rotary Vane Vacuum Pumps
575.00 SEK - 5330.00 SEK
Product Code | Brand | For Use With | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Type | Price | Quantity & Availability | ||||
10292551
|
VACUUBRAND™
20698003 |
Model RE2, RZ2, RE2.5, RZ2.5, RE5, RZ5, RE6, RZ6 rotary vane vacuum pump | Oil Mist Filter |
4515.00 SEK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
10590362
|
VACUUBRAND™
20698004 |
RZ 2.5 and RZ 6 Oil Mist Filter (20698003) | Filter Element for Oil Mist Filter |
575.00 SEK
Each |
In Stock
Log in to see stock. |
||||
10485823
|
VACUUBRAND™
20698017 |
Model RE8, RE9, RE16 and RZ8, RZ9, RZ16 rotary vane vacuum pump | Oil Mist Filter |
5330.00 SEK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
Description
For use with VACUUBRAND Rotary Vane Vacuum Pumps
- Oil Mist Filters “FO” trap up to 99% of oil mist generated by the pump, helping preserve the laboratory environment
- Cat. No. 13-889-111 is a replacement part for small oil mist filter 13-889-110