Learn More
BRD9 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24785-1-AP-20UL
 View more versions of this product
Product Code. 16883275
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The BRD9 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human BRD9. This antibody recognizes human antigen. The BRD9 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
BRD9
Polyclonal
Unconjugated
20 μL
65980
IgG
Western Blot, ELISA
587 μg/mL
Rabbit
Primary
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRD9 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 20 μL