missing translation for 'onlineSavingsMsg'
Learn More
Bromophenol blue sultone Form, MP Biomedicals Product Code.: 11308729

Bromophenol blue sultone Form, MP Biomedicals

Product Code. p-4404310 Shop All MP Biomedical Products
5 g, 5g
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11308729

Brand: MP Biomedicals 0210113005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 76-59-5
Molecular Formula C27H28Br2O5S
Molecular Weight (g/mol) 624.384
InChI Key NUHCTOLBWMJMLX-UHFFFAOYSA-N
Synonym 3,3',5,5'-Tetrabromophenolsulfonphthalein, Bromothymol Blue
PubChem CID 6450
ChEBI CHEBI:86155
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methyl-6-propan-2-ylphenol
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)C(C)C)C(C)C)O)Br

Specifications

Formula Weight 624.43
Grade ACS Reagent
Color Lilac
Quantity 5 g
Physical Form Powder
Chemical Name or Material Bromophenol blue sultone form
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromophenol blue sultone Form, MP Biomedicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.