missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BST2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13560-1-AP-20UL
This item is not returnable.
View return policy
Description
Bone marrow stromal cells are involved in the growth and development of B-cells. The specific function of the protein encoded by the bone marrow stromal cell antigen 2 is undetermined; however, this protein may play a role in pre-B-cell growth and in rheumatoid arthritis.Specifications
BST2 | |
Polyclonal | |
Unconjugated | |
BST2 | |
Bone marrow stromal antigen 2, BST 2, BST2, CD317, HM1.24 antigen, Tetherin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
684 | |
-20°C | |
Liquid |
Immunocytochemistry, Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q10589 | |
BST2 | |
BST2 Fusion Protein Ag4430 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BST2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction