missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | BTG4 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890055
|
Proteintech
20514-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880055
|
Proteintech
20514-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Shows marked antiproliferative activity, being able to induce G(1) arrest.Specifications
BTG4 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O70552, Q9NY30 | |
315650, 54766, 56057 | |
BTG4 Fusion Protein Ag14339 | |
Primary | |
-20°C | |
BTG4 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B cell translocation gene 4, BTG family member 4, BTG4, PC3B, Protein BTG4, Protein PC3b | |
Btg4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BTG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech