Learn More
BTN3A3 Mouse anti-Human, Unconjugated, Clone: 1A3B5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67560-1-IG-150UL
 View more versions of this product
Product Code. 16890256
  • 4652.90 SEK / 150µL
Description
The BTN3A3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BTN3A3. This antibody recognizes human antigen. The BTN3A3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
BTN3A3
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1A3B5
Unconjugated
150 μL
10384
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BTN3A3 Mouse anti-Human, Unconjugated, Clone: 1A3B5, Proteintech > 150 μL