missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10123-2-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
BUB3 | |
Polyclonal | |
Unconjugated | |
BUB3 | |
BUB3, BUB3L, hBUB3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12237, 361662, 9184 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43684, Q9WVA3 | |
Bub3 | |
BUB3 Fusion Protein Ag0167 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |