missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1355.00 SEK - 3665.00 SEK
Specifications
Antigen | BUB3 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16826303
|
Proteintech
10123-2-AP-20UL |
20 μL |
1355.00 SEK
20µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16816303
|
Proteintech
10123-2-AP-150UL |
150 μL |
3665.00 SEK
150µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
BUB3 | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O43684, Q9WVA3 | |
12237, 361662, 9184 | |
BUB3 Fusion Protein Ag0167 | |
Primary | |
-20°C | |
BUB3 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BUB3, BUB3L, hBUB3 | |
Bub3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |