missing translation for 'onlineSavingsMsg'
Learn More
Buffer Solution Ammonia-Ammonium Chloride, Pure, for Water Hardness Testing, Fisher Chemical™ Product Code.: 11438982

Buffer Solution Ammonia-Ammonium Chloride, Pure, for Water Hardness Testing, Fisher Chemical™

Product Code. p-7073109 Shop All Fisher Chemical Products
1 L, 1 L
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 11438982

Brand: Fisher Chemical J/2500/PB15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Buffer Solution Ammonia-Ammonium Chloride, Pure, for Water Hardness Testing

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 134124

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 L
Packaging HDPE plastic bottle
Vapor Pressure 330mmHg at 20°C
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Buffer solution ammonia-ammonium chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/A§.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution Ammonia-Ammonium Chloride, Pure, for Water Hardness Testing, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.