Learn More
Fisher Chemical Buffer Solution, Colour Coded Red, pH 4, Traceable to NIST, 1L
Brand: Fisher Chemical J/2826/15
309.71 SEK valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Buffer Solution, Colour Coded Red, pH 4, Traceable to NIST, 1L
Description
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%