Learn More
uvex Busingess Blue PU 84262 Boot - Width 11
Uvex 1 shoes designed for classic business look while providing protection against workplace risks; extra-wide steel toecap, penetration-resistant midsole and SRC polyurethane outsole protects toes and keeps from slipping; ideal for active men in office and factory environments
1570.00 SEK
Specifications
Color | Blue |
---|---|
Material | Microvelour |
Sole Material | Polyurethane (PU) |
Toe Material | Steel |
Disposable | Reusable |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17284451
|
uvex
8426239 |
39 | N/A | ||||||
17294451
|
uvex
8426240 |
40 | N/A | ||||||
17204461
|
uvex
8426241 |
41 | N/A | ||||||
17214461
|
uvex
8426242 |
42 | N/A | ||||||
17224461
|
uvex
8426243 |
43 | N/A | ||||||
17234461
|
uvex
8426244 |
44 | N/A | ||||||
17244461
|
uvex
8426245 |
45 | N/A | ||||||
17254461
|
uvex
8426246 |
46 | N/A | ||||||
17264461
|
uvex
8426247 |
47 | N/A | ||||||
17274461
|
uvex
8426248 |
48 | N/A | ||||||
17284461
|
uvex
8426249 |
49 | N/A | ||||||
17294461
|
uvex
8426250 |
50 | N/A | ||||||
17204471
|
uvex
8426251 |
51 | N/A | ||||||
17214471
|
uvex
8426252 |
52 | N/A | ||||||
Description
Uvex 1 business Chelsea Boot 8426.2 S3 SRC
- Safe, smart, stylish
- Fashionably business look with trendy design
- Generous fit with wide steel toe cap
- Penetration-resistant, non-metallic midsole in accordance with latest safety standards, does not restrict flexibility
- Uvex Climazone technology ensures optimum wearer comfort
- Breathable, water-resistant high-tech microvelour upper material
- Textile lining provides optimum breathability
- Made from synthetic materials, suitable for people allergic to chrome
- Shock absorbing, durable mono density polyurethane sole unit
- Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
- Sole unit of free of lacquer wetting inhibitors such as silicones, phthalates, etc.
- Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
- Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
- Article No. 8426.2
- Standard EN ISO 20345:2011 S3 SRC
- Sizes: 39 to 52
Specifications
Blue | |
Polyurethane (PU) | |
Reusable | |
Men's | |
Uvex 1 | |
No | |
SRC |
Microvelour | |
Steel | |
EN ISO 20345:2011 | |
Standard Width | |
Yes | |
S3 | |
ESD Safety Shoe |