missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Met (Cytoplasmic) Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | c-Met (Cytoplasmic) |
---|---|
Concentration | 0.29 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884425
|
Proteintech
25869-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16874425
|
Proteintech
25869-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
In Western blot, this antibody produces bands of unknown identity at 55 and 100 kDa.
The Mesenchymal Epithelial Transition (MET) receptor belongs to a family of receptor tyrosine kinases (RTKs). Dyregulation of the MET signaling pathway occurs in wide range of human tumors. The MET receptor is a glycoprotein heterodimer consisting of an extracellular alpha-subunit linked to transmembrane beta-subunit by a disulfide bond. The MET receptor is widely expressed on the surface of epithelial and endothelial cells. MET ligand is known as the hepatocyte growth factor (HGF), also called the scatter factor (SF). HGF/SF is a serine protease and produced in cells of mesenchymal origin. HGF is secreted as a s ingle chain, biologically inactive protein which is further processed into its mature form by a cleavage process catalyzed by extracellular proteases. MET and its ligand HGF are involved in several normal and pathological processes, such as fetal liver, placenta and muscle development, as well as development of the nervous system. MET signal transduction is involved in tumor growth, invasion, resistance to therapy, angiogenesis and specially in the generation and maintece of cancer stem cells (CSCs). Upon HGF binding, the MET receptor at the plasma membrane forms a stable dimer of two molecules of MET which leads to subsequent activation and trans-phosphorylation of the two tyrosine residues in the catalytical regions Y1234 and Y1235, followed by trans-phosphorylation of two docking tyrosines (Y1349 and Y1356) in the carboxy-terminal site. These phosphorylations enable MET to bind to multiple substrates and activate a variety of signaling pathways including MAPK, PI3K-AKT cascades, STAT and NF-kappaB.Specifications
c-Met (Cytoplasmic) | |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
P08581 | |
4233 | |
c-Met (Cytoplasmic) Fusion Protein Ag23140 | |
Primary | |
-20°C | |
MET |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AUTS9, c-Met, HGF receptor, HGF/SF receptor, HGFR, MET, Proto oncogene c Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine protein kinase Met | |
MET | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
c-Met (Cytoplasmic) Rabbit anti-Human, Polyclonal, Proteintech