missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf54 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23251-1-AP-20UL
This item is not returnable.
View return policy
Description
C11orf54 protein function is currently unknown.Specifications
C11orf54 | |
Polyclonal | |
Unconjugated | |
C11orf54 | |
C11orf54, Ester hydrolase C11orf54, PTD012 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
28970, 363016 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H0W9 | |
C11orf54, RGD1309534 | |
C11orf54 Fusion Protein Ag19731 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C11orf54 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction