missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf67 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | C11orf67 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899944
|
Proteintech
20414-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889944
|
Proteintech
20414-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AAMDC (C11orf67) may play a role in preadipocyte differentiation and adipogenesis.Specifications
C11orf67 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q8R0P4, Q9H7C9 | |
28971, 361606, 66273 | |
C11orf67 Fusion Protein Ag14258 | |
Primary | |
-20°C | |
AAMDC |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AAMDC, C11orf67, CK067, PTD015, UPF0366 protein C11orf67 | |
AAMDC | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C11orf67 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech