Learn More
C11orf70 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20780-1-AP-20UL
This item is not returnable.
View return policy
Specifications
C11orf70 | |
Polyclonal | |
Unconjugated | |
C11orf70 | |
CK070, C11orf70 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
85016 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BRQ4 | |
C11orf70 | |
C11orf70 Fusion Protein Ag14793 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
C11orf70 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction