missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C15orf58 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | C15orf58 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884025
|
Proteintech
25447-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16874025
|
Proteintech
25447-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specific and highly efficient GDP-D-glucose phosphorylase regulating the levels of GDP-D-glucose in cells.Specifications
C15orf58 | |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q3TLS3, Q6ZNW5 | |
269952, 390637 | |
C15orf58 Fusion Protein Ag20215 | |
Primary | |
-20°C | |
GDPGP1 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C15orf58, UPF0580 protein C15orf58 | |
GDPGP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C15orf58 Rabbit anti-Human, Mouse, Polyclonal, Proteintech