Learn More
C16orf61 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26086-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Product No. 26086-1-AP is a rabbit polyclonal antibody raised against the full-length of human C16orf61. The MW of this protein is 9 kDa, and this antibody specifically recognizes the 9 kDa protein.
The function of C16orf61 remains largely unknown. It may be involved in cytochrome c oxidase biogenesis.Specifications
C16orf61 | |
Polyclonal | |
Unconjugated | |
CMC2 | |
2310061C15Rik, C16orf61, CMC2, DC13 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56942 | |
-20°C | |
Liquid |
Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NRP2 | |
CMC2 | |
C16orf61 Fusion Protein Ag23319 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |