missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf61 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | C16orf61 |
---|---|
Concentration | 0.39 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824615
|
Proteintech
26086-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16814615
|
Proteintech
26086-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Product No. 26086-1-AP is a rabbit polyclonal antibody raised against the full-length of human C16orf61. The MW of this protein is 9 kDa, and this antibody specifically recognizes the 9 kDa protein.
The function of C16orf61 remains largely unknown. It may be involved in cytochrome c oxidase biogenesis.Specifications
C16orf61 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9NRP2 | |
56942 | |
C16orf61 Fusion Protein Ag23319 | |
Primary | |
-20°C | |
CMC2 |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
2310061C15Rik, C16orf61, CMC2, DC13 | |
CMC2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |