Learn More
C19orf12 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27382-1-AP-20UL
 View more versions of this product
Product Code. 16885595
  • 1645.54 SEK / 20µL
Specifications
C19orf12
Polyclonal
Unconjugated
C19orf12
C19orf12
Rabbit
Antigen Affinity Chromatography
RUO
83636
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9NSK7
C19orf12
C19orf12 Fusion Protein Ag26555
20 μL
Primary
Human
Antibody
IgG
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
C19orf12 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated