Learn More
C1orf103 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | C1orf103 |
---|---|
Concentration | 0.26 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884645
|
Proteintech
26115-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16874645
|
Proteintech
26115-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The specific function of this protein remains unknown.Specifications
C1orf103 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q5T3J3, Q8CDD9 | |
321000, 55791 | |
C1orf103 Fusion Protein Ag23494 | |
Primary | |
-20°C | |
LRIF1 |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C1orf103, LRIF1, Receptor interacting factor 1, RIF1, RP11 96K19.1 | |
Lrif1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
C1orf103 Rabbit anti-Human, Mouse, Polyclonal, Proteintech