missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf156 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25553-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
C1orf156 | |
Polyclonal | |
Unconjugated | |
METTL18 | |
AsTP2, C1orf156, METTL18, RP1 117P20.4, UPF0558 protein C1orf156 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
92342 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95568 | |
METTL18 | |
C1orf156 Fusion Protein Ag22148 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |