missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf156 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | C1orf156 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16884115
|
Proteintech
25553-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16874115
|
Proteintech
25553-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
C1orf156 | |
Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
O95568 | |
92342 | |
C1orf156 Fusion Protein Ag22148 | |
Primary | |
-20°C | |
METTL18 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AsTP2, C1orf156, METTL18, RP1 117P20.4, UPF0558 protein C1orf156 | |
METTL18 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C1orf156 Rabbit anti-Human, Polyclonal, Proteintech