Learn More
C1orf216 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26265-1-AP-20UL
 View more versions of this product
Product Code. 16884775
  • 1645.54 SEK / 20µL
Description
C1orf216 is a protein coding gene.
Specifications
Specifications
C1orf216
Polyclonal
Unconjugated
C1orf216
C1orf216, UPF0500 protein C1orf216
Rabbit
Antigen Affinity Chromatography
RUO
127703
-20°C
Liquid
Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8TAB5
C1orf216
C1orf216 Fusion Protein Ag22665
20 μL
Primary
Mouse, Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
C1orf216 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated