Learn More
C1orf27 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22844-1-AP-20UL
 View more versions of this product
Product Code. 16882105
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
May play a role in the trafficking of a subset of G-protein coupled receptors.
Specifications
Specifications
C1orf27
Polyclonal
Unconjugated
ODR4
C1orf27, hODR 4, LAG1 interacting protein, odr 4, ODR4, Protein odr 4 homolog, TTG1, TTG1A
Rabbit
Antigen Affinity Chromatography
RUO
226499, 54953
-20°C
Liquid
Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4PJX1, Q5SWX8
ODR4
C1orf27 Fusion Protein Ag18802
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C1orf27 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated