Learn More
C1QA Mouse anti-Human, Unconjugated, Clone: 1B10E3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67063-1-IG-150UL
 View more versions of this product
Product Code. 16879465
  • 4652.90 SEK / 150µL
Description
The C1QA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human C1QA. This antibody recognizes human antigen. The C1QA antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
C1QA
Monoclonal
500 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1B10E3
Unconjugated
150 μL
712
IgG1
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
C1QA Mouse anti-Human, Unconjugated, Clone: 1B10E3, Proteintech > 150 μL