missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24474-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes one of two duplicated zinc finger genes on chromosome Xp11. This gene is the telomeric copy; GeneID 158586 ZXDB is the more centromeric copy. The two genes have 98% nucleotide sequence similarity, and the predicted proteins contain 10 tandem zinc finger motifs.Specifications
C1QBP | |
Polyclonal | |
Unconjugated | |
C1QBP | |
C1QBP, gC1Q R, GC1q R protein, GC1QBP, gC1qR, Glycoprotein gC1qBP, HABP1, Hyaluro binding protein 1, p32, p33, SF2p32 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12261, 708 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35658, Q07021 | |
C1QBP | |
C1QBP Fusion Protein Ag19773 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C1QBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction