missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1qC Mouse anti-Human, Clone: 1C5B4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66268-1-IG-20UL
This item is not returnable.
View return policy
Description
This antibody is raised against C1qC which is the C-chain polypeptide of human complement subcomponent C1q.
This gene encodes a major constituent of the human complement subcomponent C1q. C1q associates with C1r and C1s in order to yield the first component of the serum complement system. A deficiency in C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N-terminus, and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1. This gene encodes the C-chain polypeptide of human complement subcomponent C1q. Alternatively spliced transcript variants that encode the same protein have been found for this gene.Specifications
C1qC | |
Monoclonal | |
0.68 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02747 | |
C1QC | |
C1qC Fusion Protein Ag10233 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgM |
Western Blot | |
1C5B4 | |
Unconjugated | |
C1QC | |
C1Q C, C1qC, C1QG | |
Mouse | |
Affinity Chromatography | |
RUO | |
714 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C1qC Mouse anti-Human, Clone: 1C5B4, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction