Learn More
C1QTNF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11414-1-AP-20UL
 View more versions of this product
Product Code. 16888473
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Multiple isoforms of human CTRP3 have been reported. It has been suggested that CTRP3 may play a role in skeletal development.
Specifications
Specifications
C1QTNF3
Polyclonal
Unconjugated
C1QTNF3
C1ATNF3, C1QTNF3, Corcs, Cors, CORS26, CTRP3, Secretory protein CORS26
Rabbit
Antigen Affinity Chromatography
RUO
114899, 294806, 81799
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BXJ4, Q9ES30
C1qtnf3
C1QTNF3 Fusion Protein Ag1958
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C1QTNF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated