Learn More
C20orf43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16633-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Product No. 16633-1-AP is a rabbit polyclonal antibody raised against the full-length of human C20orf43. The MW of this protein is 40 kDa, and this antibody specifically recognizes the 40 kDa protein.
C20orf43 protein function remains unknown.Specifications
C20orf43 | |
Polyclonal | |
Unconjugated | |
RTFDC1 | |
C20orf43, CDA05, CDAO5, HSPC164, HSPC169, RTF2, SHUJUN 3, UPF0549 protein C20orf43 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
296410, 51507, 66404 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q3T1J8, Q99K95, Q9BY42 | |
Rtfdc1 | |
C20orf43 Fusion Protein Ag9972 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |