missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf43 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | C20orf43 |
---|---|
Concentration | 0.43 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836314
|
Proteintech
16633-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826314
|
Proteintech
16633-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Product No. 16633-1-AP is a rabbit polyclonal antibody raised against the full-length of human C20orf43. The MW of this protein is 40 kDa, and this antibody specifically recognizes the 40 kDa protein.
C20orf43 protein function remains unknown.Specifications
C20orf43 | |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q3T1J8, Q99K95, Q9BY42 | |
296410, 51507, 66404 | |
C20orf43 Fusion Protein Ag9972 | |
Primary | |
-20°C | |
RTFDC1 |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
C20orf43, CDA05, CDAO5, HSPC164, HSPC169, RTF2, SHUJUN 3, UPF0549 protein C20orf43 | |
Rtfdc1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |