missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf46 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24678-1-AP-20UL
This item is not returnable.
View return policy
Description
C20orf46, also named as TMEM74B, belongs to the TMEM74 family.Specifications
C20orf46 | |
Polyclonal | |
Unconjugated | |
TMEM74B | |
C20orf46, Transmembrane protein C20orf46 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108832, 55321 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NUR3 | |
TMEM74B | |
C20orf46 Fusion Protein Ag20385 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C20orf46 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction