Learn More
C2orf49 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18272-1-AP-150UL
 View more versions of this product
Product Code. 16888334
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The function of this protein has not been specifically defined.
Specifications
Specifications
C2orf49
Polyclonal
Unconjugated
C2orf49
Ashwin, C2orf49
Rabbit
Antigen Affinity Chromatography
RUO
79074
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BVC5
C2orf49
C2orf49 Fusion Protein Ag12948
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C2orf49 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated