Learn More
C5orf24 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24319-1-AP-20UL
This item is not returnable.
View return policy
Description
C5orf24 is a protein coding gene.Specifications
C5orf24 | |
Polyclonal | |
Unconjugated | |
C5orf24 | |
C5orf24, UPF0461 protein C5orf24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
134553 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7Z6I8 | |
C5orf24 | |
C5orf24 Fusion Protein Ag19534 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
C5orf24 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction