Learn More
C5orf26 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24698-1-AP-20UL
This item is not returnable.
View return policy
Specifications
C5orf26 | |
Polyclonal | |
Unconjugated | |
EPB41L4A-AS1 | |
EPB41L4A-AS1 | |
C5orf26 Fusion Protein Ag20367 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C5orf26, TIGA1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114915 | |
-20°C | |
Liquid |
Product Content Correction
Product Title
C5orf26 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction