missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C6orf15 Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24953-1-AP-20UL
This item is not returnable.
View return policy
Description
C6orf15, also named as Protein STG, is a 325 amino acid protein, which as a secreted protein is expressed in skin and tonsils. The calculated molecular weight of C6orf15 is 34 kDa, but we detected a 50-55 kDa protein by western blot. C6orf15 may contains some modified residues.Specifications
C6orf15 | |
Polyclonal | |
Unconjugated | |
C6orf15 | |
C6orf15, Protein STG, STG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29113 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6UXA7 | |
C6orf15 | |
C6orf15 Fusion Protein Ag20276 | |
20 ÎĽL | |
Primary | |
Non-human Primate, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C6orf15 Rabbit anti-Human, Non-human primate, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction