missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf41 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24068-1-AP-20UL
This item is not returnable.
View return policy
Description
N-methyltransferase that mediates the formation of anserine (beta-alanyl-N(Pi)-methyl-L-histidine) from carnosine. Anserine, a methylated derivative of carnosine (beta-alanyl-L-histidine), is an abundant constituent of vertebrate skeletal muscles. Also methylates other L-histidine-containing di- and tripeptides such as Gly-Gly-His, Gly-His and homocarnosine (GABA-His).Specifications
C9orf41 | |
Polyclonal | |
Unconjugated | |
CARNMT1 | |
C9orf41, UPF0586 protein C9orf41 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
138199 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N4J0 | |
CARNMT1 | |
C9orf41 Fusion Protein Ag21268 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C9orf41 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction