Learn More
C9orf46 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26101-1-AP-20UL
 View more versions of this product
Product Code. 16884625
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Specifications
C9orf46
Polyclonal
Unconjugated
PLGRKT
AD025, C9orf46, MDS030, Transmembrane protein C9orf46
Rabbit
Antigen Affinity Chromatography
RUO
55848
-20°C
Liquid
Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9HBL7
PLGRKT
C9orf46 Fusion Protein Ag23410
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
C9orf46 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated