Learn More
C9orf72 Mouse, anti-Human, Unconjugated, Clone: 2F9A2, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | C9orf72 |
---|---|
Clone | 2F9A2 |
Concentration | 1000 ÎĽg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17231476
|
Proteintech
67824-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17221476
|
Proteintech
67824-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C9ORF72 has a domain whith polymorphic hexanucleotide repeat (GGGGCC). The C9ORF72-hexanucleotide repeat expansions have been recently identified as genetic markers in amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD). The C9ORF72 repeat expansions may indicate a worse prognosis in ALS. Human C9ORF72 has some isoforms with MW 54-60 kDa and 25-30 kDa. Mouse C9orf72 has some isoforms with MW 50-60 kDa and 35 kDa.
Specifications
C9orf72 | |
1000 ÎĽg/mL | |
Monoclonal | |
Liquid | |
Human, Mouse, Rat, Pig | |
Primary |
2F9A2 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
203228 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.