missing translation for 'onlineSavingsMsg'
Learn More

CACNG2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20969-1-AP-150UL

 View more versions of this product

Product Code. 16880555

  • 4652.90 SEK / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Stargazin is a member of the transmembrane AMPAR regulatory proteins (TARP) family and is involved in glutamate receptor trafficking. It has been recently demonstrated that the interaction between stargazin and AMPA receptors is critical for the correct localization of the receptors at the synapse. Phosphorylation of the stargazin protein at Thr 321 by Protein Kinase A regulates its interaction with PSD-95 and synaptic targeting of AMPA receptors.
Specifications

Specifications

CACNG2
Polyclonal
Unconjugated
CACNG2
CACNG2, stargazin, TARP gamma-2
Rabbit
Antigen Affinity Chromatography
RUO
10369, 12300
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88602, Q9Y698
CACNG2
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CACNG2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.