missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16617-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The de novo synthesis of pyrimidine nucleotides is required for mammalian cells to proliferate. This gene encodes a trifunctional protein which is associated with the enzymatic activities of the first 3 enzymes in the 6-step pathway of pyrimidine biosynthesis: carbamoylphosphate synthetase (CPS II), aspartate transcarbamoylase, and dihydroorotase. This protein is regulated by the mitogen-activated protein kinase (MAPK) cascade, which indicates a direct link between activation of the MAPK cascade and de novo biosynthesis of pyrimidine nucleotides.Specifications
CAD | |
Polyclonal | |
Unconjugated | |
CAD | |
CAD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
790 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunofluorescence, Immunocytochemistry | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P27708 | |
CAD | |
CAD Fusion Protein Ag9912 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |