missing translation for 'onlineSavingsMsg'
Learn More
Cafestol, 98.92%, For HPLC analysis, MP Biomedicals™ Product Code.: 11432321

Cafestol, 98.92%, For HPLC analysis, MP Biomedicals™

Product Code. 11432321 Shop All MP Biomedical Products
100 mg
Click to view available options
Quantity:
50 mg
100 mg
This item is not returnable. View return policy

Product Code. 11432321

Brand: MP Biomedicals 0219374580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cafestol is a diterpene molecule present in coffee

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 469-83-0
Molecular Formula C20H28O3
Molecular Weight (g/mol) 316.44
MDL Number MFCD01075769
InChI Key DNJVYWXIDISQRD-HWUKTEKMSA-N
Synonym cafestol, cafesterol, ccris 1518, unii-ac465t6q6w, 5a,8-methano-5ah-cyclohepta 5,6 naphtho 2,1-b furan-7-methanol, 3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-7-hydroxy-10b-methyl-, 3bs,5as,7r,8r,10ar,10bs, 5a,8-methano-5ah-cyclohepta 5,6 naphtho 2,1-b furan-7-methanol, 3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-7-hydroxy-10b-methyl-, 3bs-3balpha,5abeta,7beta,8beta,10aalpha,10bbeta, --cafestol, 1s,4s,12s,13r,16r,17r-17-hydroxymethyl-12-methyl-8-oxapentacyclo 14.2.1.0 1,13 .0 4,12 .0 5,9 nonadeca-5 9 ,6-dien-17-ol, 3bs,5as,7r,8r,10ar,10bs-7-hydroxymethyl-10b-methyl-3b,4,5,6,7,8,9,10,10a,10b,11,12-dodecahydro-5a,8-methanocyclohepta 5,6 naphtho 2,1-b furan-7-ol
PubChem CID 108052
ChEBI CHEBI:3291
IUPAC Name (1S,4S,12S,13R,16R,17R)-17-(hydroxymethyl)-12-methyl-8-oxapentacyclo[14.2.1.0¹,¹³.0⁴,¹².0⁵,⁹]nonadeca-5(9),6-dien-17-ol
SMILES C[C@@]12CCC3=C(C=CO3)[C@H]1CC[C@@]13C[C@@H](CC[C@@H]21)[C@@](O)(CO)C3

Specifications

Color White
Quantity 100 mg
Formula Weight 316.4
Physical Form Powder
Chemical Name or Material Cafestol, For HPLC analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cafestol, 98.92%, For HPLC analysis, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.