missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sensing Receptor Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19125-1-AP-150UL
This item is not returnable.
View return policy
Description
The 60-65 kDa band detected by this antibody might represent a degradation product of CASR.
Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.Specifications
Calcium Sensing Receptor | |
Polyclonal | |
Unconjugated | |
CASR | |
Calcium Sensing Receptor, CAR, CASR, FHH, FIH, GPRC2A, HHC, HHC1, NSHPT, PCAR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12374, 24247, 846 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P41180, P48442, Q9QY96 | |
CASR | |
Peptide | |
150 ÎĽL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Calcium Sensing Receptor Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction