Learn More
Calibration Buffer Solutions, Hanna Instruments™
Click to view available options
Quantity:
20 mL
230 mL
1 L
pH:
4.01
9.18
10.01
Unit Size:
Each
Pack of 25
Description
- Premium quality pH 10.01 calibration buffer solution
- 25 sachets containing 20 mL of buffer
- Color coded label for easy pH buffer value identification
- Accuracy of +/- 0.01 pH @ 25°C
- Temperature chart of the actual pH value at various temperatures printed on each sachet
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware
- In a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
- Ideal for users that do not calibrate frequently
- Ideal for field use due to portability
Specifications
Specifications
Color | Colorless |
pH | 9.18 |
Physical Form | Liquid |
Quantity | 230 mL |
Solution Type | Buffer |
Formulation | Aqueous Buffer Solution |
Odor | Odorless |
Product Content Correction
Product Title
Calibration Buffer Solutions, Hanna Instruments™ >
Spot an opportunity for improvement?Share a Content Correction