Learn More
CalPlus™ (Brand of Calcium Chloride Dihydrate), Multi-Compendial, U.S.P., J.T. Baker™ Product Code.: 15579400
compliance-icons
CalPlus™ (Brand of Calcium Chloride Dihydrate), Multi-Compendial, U.S.P., J.T. Baker™
Product Code. p-5390732
500 g, Wide Mouth Amber Glass Bottle, 500g
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle
Unit Size:
2.50kg
500g
Product Code. 15579400
Brand: Avantor J.T.Baker 133601
Suitable for Use in Hemodialysis
Chemical Identifiers
CAS 10035-04-8
Molecular Formula CaCl2·2H2O
Specifications
Quantity 500 g
Formula Weight 147.01
Grade Multi-Compendial/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 0.84kg
Chemical Name or Material CalPlus™ (Brand of Calcium Chloride Dihydrate)
compliance-icons
Product Identifier
  • Calcium Chloride Dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Calcium Chloride Dihydrate
Product Content Correction
Product Title
CalPlus™ (Brand of Calcium Chloride Dihydrate), Multi-Compendial, U.S.P., J.T. Baker™ >