missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CAMK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13730-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody 13730-1-AP recognizes all the members of CAMK2 family.
The Ca2+/calmodulin-dependent protein kinases (CaM kinases) comprise a structurally related subfamily of serine/threonine kinases which include CaMKI, CaMKII and CaMKIV. CaMKII is an ubiquitously expressed serine/threonine protein kinase that is activated by Ca2+/calmodulin (CaM) binding and autophosphorylation and has been implicated in regulation of the cell cycle and transcription. It is involved in dendritic spine and synapse formation, neuronal plasticity and regulation of sarcoplasmic reticulum Ca2+ transport in skeletal muscle. In neurons, plays an essential structural role in the reorganization of the actin cytoskeleton during plasticity by binding and bundling actin filaments in a kinase-independent manner.Specifications
CAMK2 | |
Polyclonal | |
Unconjugated | |
CAMK2A | |
CaM kinase II subunit alpha, CaMK II subunit alpha, CAMK2, CAMK2A, CAMK2B, CAMK2D, CAMK2G, CAMKA, CaMKII alpha, KIAA0968 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108058, 12322, 12323, 12325, 171140, 24245, 24246, 25400, 815, 816, 817, 818 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P28652, Q13554, Q13555, Q9UQM7 | |
CAMK2A, CAMK2B, CAMK2D, Camk2g | |
CAMK2 Fusion Protein Ag4745 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CAMK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction