missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CAMK2B (CaMKII Beta), His Tag Recombinant Protein
Human CAMK2B (CaMKII Beta), His Tag Recombinant Protein
Brand: Thermo Scientific™ PR7771A
Additional Details : Weight : 0.01000kg
Description
As a dominant kinase in the central nervous system CAMK2B (CaMKII beta) may function in long term potentiation and neurotransmitter release.Specifications
NP_742078 | |
60.5 kDa | |
CAMK2B (CaMKII Beta), CAMK2 | |
CAMK2B Recombinant Human Protein | |
Recombinant | |
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, and 50% Glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
Kinase Assay | |
100 μg | |
CAMK2 Kinase Family | |
Human | |
His Tag | |
Ser⁄Thr Kinases: CAMK Group | |
Baculovirus | |
Recombinant, Full Length | |
Serine/Threonine Kinases |